Supelco® PESTANAL® Organic Reference Standard, Orthosulfamuron
Catalog # 32427-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:213464-77-8
- Description:Orthosulfamuron
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD09027340
- M-Clarity quality level:MQ100
- Synonym(s):1-(4,6-Dimethoxy-2-pyrimidinyl)-3-[2-(dimethylcarbamoyl)phenylsulfamoyl]urea
- Formula:C₁₆H₂₀N₆O₆S
- Molecular weight:424,43
- Flash point:Not applicable