Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Octocrylene, analytical standard, Supelco®
Organic reference standard, Octocrylene, analytical standard, Supelco®
Catalog # 02343-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Octocrylene, analytical standard, Supelco®
Catalog # 02343-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    6197-30-4
  • Description:
    Octocrylene
  • Melting point:
    –10 °C (lit.)
  • Formula:
    C₂₄H₂₇NO₂
  • Description:
    Octocrylene
  • Boiling point:
    218 °C/1.5 mm Hg (lit.)

Specifications

About this item