Supelco® PESTANAL® Organic Reference Standard, Methoxyfenozide
Catalog # 32507-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:161050-58-4
- Description:Methoxyfenozide
- Storage conditions:2...30 °C
- Hazard class description:H312 - H411
- Shipping conditions:STD
- MDL No.:MFCD03792750
- M-Clarity quality level:MQ100
- Synonym(s):3-Methoxy-2-methylbenzoic acid 2-(3,5-dimethylbenzoyl)-2-tert-butylhydrazide
- Formula:C₂₂H₂₈N₂O₃
- Molecular weight:368,47
- Flash point:Not applicable
Specifications
About this item
Caution: P273 - P280 - P302 + P352 + P312 - P362 + P364 - P391 - P501