Supelco® PESTANAL® Organic Reference Standard, 3,6-Dihydroxyphthalonitrile
Catalog # 36611-1G-R
Supplier: Merck
CAS Number:
Specifications
- Pk:1 g
- Standards:PESTANAL®, Analytical standard
- CAS No.:4733-50-0
- Description:3,6-Dihydroxyphthalonitrile
- Storage conditions:2...8 °C
- Shipping conditions:STD
- MDL No.:MFCD00001790
- M-Clarity quality level:MQ100
- Synonym(s):2,3-Dicyano-1,4-hydroquinone; 2,3-Dicyanohydroquinone; DCH
- Melting point:190...196 °C (subl,) (lit.)
- Formula:C₈H₁₆O₄
- Molecular weight:160,13
- Storage temperature:2…8 °C
- Flash point:55 °C - closed cup
Specifications
About this item
- GHS signal: GHS02, GHS06