Order Entry
Austria
ContactUsLinkComponent
Pesticide reference standard, Isoxathion, analytical standard, Supelco®
Pesticide reference standard, Isoxathion, analytical standard, Supelco®
Catalog # 76529-25MG
Supplier:  Merck
CAS Number:  
undefined
Pesticide reference standard, Isoxathion, analytical standard, Supelco®
Catalog # 76529-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    Analytical standard
  • CAS No.:
    18854-01-8
  • Description:
    Isoxathion
  • Formula:
    C₁₃H₁₆NO₄PS
  • Description:
    Isoxathion

Specifications

About this item

Packaging: Bottomless glass bottles are inserted with fused cone.