Supelco® PESTANAL® Organic Reference Standard, Iminoctadine Triacetate
37007-10MG
: Merck
:
- Pk:10 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:57520-17-9
- Description:Iminoctadine triacetate
- Storage conditions:2...8 °C
- Hazard class description:H301 - H312 - H411
- Shipping conditions:WICE
- MDL No.:MFCD01724078
- M-Clarity quality level:MQ100
- Synonym(s):N,N″′-(Iminodi-8,1-octanediyl)bisguanidine triacetate
- Formula:C₂₄H₅₃N₇O₆
- Molecular weight:535,72
- Storage temperature:2...8 °C
- Flash point:Not applicable
- GHS signal: GHS06, GHS09
: P273 - P280 - P301 + P310 + P330 - P302 + P352 + P312