Order Entry
Austria
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Imazethapyr
Supelco® PESTANAL® Organic Reference Standard, Imazethapyr
Catalog # 37923-100MG-R
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Imazethapyr
Catalog # 37923-100MG-R
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    81335-77-5
  • Description:
    Imazethapyr
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00274561
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    2-(4,5-Dihydro-4-isopropyl-4-methyl-5-oxo-1H-imidazol-2-yl)-5-ethyl-3-pyridinecarboxylic acid
  • Formula:
    C₁₅H₁₉N₃O₃
  • Molecular weight:
    289,33
  • Flash point:
    Not applicable

Specifications

About this item