Supelco® PESTANAL® Organic Reference Standard, Furalaxyl
Catalog # 45516-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:57646-30-7
- Description:Furalaxyl
- Storage conditions:2...30 °C
- Hazard class description:H302 - H412
- Shipping conditions:STD
- MDL No.:MFCD00078674
- M-Clarity quality level:MQ100
- Formula:C₁₇H₁₉NO₄
- Molecular weight:301,34
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS07
Caution: P273 - P301 + P312 + P330