Supelco® PESTANAL® Organic Reference Standard, Flutriafol
Catalog # 34344-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:76674-21-0
- Description:Flutriafol
- Storage conditions:2...30 °C
- Hazard class description:H302 - H411
- Shipping conditions:STD
- MDL No.:MFCD00144319
- M-Clarity quality level:MQ100
- Formula:C₁₆H₁₃F₂N₃O
- Molecular weight:301,29
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS07
Caution: P264 - P270 - P273 - P301 + P312 - P391 - P501