Supelco® PESTANAL® Organic Reference Standard, Flucycloxuron
Catalog # 32529-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:PESTANAL®, Analytical standard, mixture of (E) and (Z) isomers
- CAS No.:113036-88-7
- Description:Flucycloxuron
- Storage conditions:2...30 °C
- Hazard class description:H319 - H413
- Shipping conditions:STD
- MDL No.:MFCD01632755
- M-Clarity quality level:MQ100
- Synonym(s):N-[[[4-[[[[(4-Chlorophenyl)cyclopropylmethylene]amino]oxy]methyl]phenyl]amino]carbonyl]-2,6-difluorobenzamide
- Formula:C₂₅H₂₀ClF₂N₃O₃
- Molecular weight:483,89
Specifications
About this item
- GHS signal: GHS07
Caution: P264 - P273 - P280 - P305 + P351 + P338 - P337 + P313 - P501