Supelco® PESTANAL® Organic Reference Standard, Flonicamid
32509-25MG
: Merck
:
- Pk:25 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:158062-67-0
- Description:Flonicamid
- Storage conditions:2...30 °C
- Hazard class description:H302
- Shipping conditions:STD
- MDL No.:MFCD07364026
- M-Clarity quality level:MQ100
- Synonym(s):N-(Cyanomethyl)-4-(trifluoromethyl)nicotinamide; N-Cyanomethyl-4-trifluoromethyl-3-pyridinecarboxamide
- Formula:C₉H₆F₃N₃O
- Molecular weight:229,16
- Flash point:Not applicable
- GHS signal: GHS07
: P264 - P270 - P301 + P312 - P501