Order Entry
Austria
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, Fenamiphos
  45483-250MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Fenamiphos
  45483-250MG
 :  Merck
 :  

 

  • Pk:
    250 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    22224-92-6
  • Description:
    Fenamiphos
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H300 + H310 + H330 - H319 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00055454
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Phenamiphos
  • Formula:
    C₁₃H₂₂NO₃PS
  • Molecular weight:
    303,36
  • Storage temperature:
    2...8 °C
  • Flash point:
    Not applicable

 

 

  • GHS signal: GHS06, GHS09
 : P262 - P273 - P280 - P302 + P352 + P310 - P304 + P340 + P310 - P305 + P351 + P338