Order Entry
Austria
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Etaconazol
Supelco® PESTANAL® Organic Reference Standard, Etaconazol
Catalog # 45471-250MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Etaconazol
Catalog # 45471-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    60207-93-4
  • Description:
    Etaconazol
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00078700
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₁₄H₁₅Cl₂N₃O₂
  • Molecular weight:
    328,19
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS07
Caution: P264 - P270 - P301 + P312 - P501