Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Dodecamethylcyclohexasiloxane, analytical standard, Supelco®
Catalog # 43216-25MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Dodecamethylcyclohexasiloxane, analytical standard, Supelco®
Catalog # 43216-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    Analytical standard
  • CAS No.:
    540-97-6
  • Description:
    Dodecamethylcyclohexasiloxane
  • Formula:
    C₁₂H₃₆O₆Si₆
  • Description:
    Dodecamethylcyclohexasiloxane

Specifications

About this item