Organic reference standard, Dioxybenzone, analytical standard, Supelco®
92841-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:131-53-3
- Description:Dioxybenzone
- Melting point:72…74 °C; 73...75 °C (lit.)
- Formula:C₁₄H₁₂O₄
- Description:Dioxybenzone
- Boiling point:170...175 °C/1 mmHg(lit.)
: Bottomless glass bottles are inserted with fused cone.