Order Entry
Austria
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Dimethoate
Supelco® PESTANAL® Organic Reference Standard, Dimethoate
  45449-100MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Dimethoate
  45449-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    60-51-5
  • Description:
    Dimethoate
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H301 + H311
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00053676
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₅H₁₂NO₃PS₂
  • Molecular weight:
    229,26
  • Storage temperature:
    2...8 °C
  • Flash point:
    107 °C - closed cup

 

 

  • GHS signal: GHS07
 : P264 - P280 - P301 + P310 - P302 + P352 + P312 - P361 + P364 - P405