Organic reference standard, Decamethylcyclopentasiloxane, analytical standard, Supelco®
43217-250MG
: Merck
:
- Pk:250 mg
- Standards:Analytical standard
- CAS No.:541-02-6
- Description:Decamethylcyclopentasiloxane
- Formula:C₁₀H₃₀O₅Si₅
- Description:Decamethylcyclopentasiloxane
- Boiling point:90 °C/10 mmHg(lit.)