Order Entry
Austria
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Clothianidin
Supelco® PESTANAL® Organic Reference Standard, Clothianidin
Catalog # 33589-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Clothianidin
Catalog # 33589-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    210880-92-5
  • Description:
    Clothianidin
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD06200753
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (E)-1-(2-Chloro-5-thiazolylmethyl)-3-methyl-2-nitroguanidine
  • Formula:
    C₆H₈ClN₅O₂S
  • Molecular weight:
    249,68
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS07, GHS09
Caution: P264 - P270 - P273 - P301 + P312 - P391 - P501