Supelco® PESTANAL® Organic Reference Standard, Chlorothalonil
Catalog # 36791-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:1897-45-6
- Description:Chlorothalonil
- Storage conditions:2...30 °C
- Hazard class description:H317 - H318 - H330 - H335 - H351 - H410
- Shipping conditions:STD
- MDL No.:MFCD00045594
- M-Clarity quality level:MQ100
- Synonym(s):Tetrachloroisophthalodinitrile
- Formula:C₈Cl₄N₂
- Molecular weight:265,91
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS06, GHS08, GHS05, GHS09
Caution: P202 - P273 - P280 - P302 + P352 - P304 + P340 + P310 - P305 + P351 + P338