Supelco® PESTANAL® Organic Reference Standard, Baythroid®
33738-250MG
: Merck
:
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:68359-37-5
- Description:Baythroid®
- Storage conditions:2...30 °C
- Hazard class description:H300 - H331 - H410
- Shipping conditions:STD
- MDL No.:MFCD00078636
- M-Clarity quality level:MQ100
- Synonym(s):Cyfluthrin
- Formula:C₂₂H₁₈Cl₂FNO₃
- Molecular weight:434,29
- Flash point:Not applicable
- GHS signal: GHS06, GHS09
: P261 - P264 - P270 - P273 - P301 + P310 - P304 + P340 + P311