Order Entry
Austria
ContactUsLinkComponent
Supelco® Organic Reference Standard, 4-(3,6-Dimethyl-3-Heptyl)Phenol-Ring-¹³C₆ Solution
Supelco® Organic Reference Standard, 4-(3,6-Dimethyl-3-Heptyl)Phenol-Ring-¹³C₆ Solution
Catalog # 33574-1ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, 4-(3,6-Dimethyl-3-Heptyl)Phenol-Ring-¹³C₆ Solution
Catalog # 33574-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    1173020-38-6
  • Description:
    4-(3,6-Dimethyl-3-heptyl)phenol-ring-¹³C₆ solution
  • Seq:
    01
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H225 - H319 - H336 - H361
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD09842832
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    3,6,3-Nonylphenol-¹³C₆; 363-NP-13C6; 4-(1-Ethyl-1,4-dimethylpentyl)phenol-¹³C₆ (ring-¹³C₆)
  • Formula:
    ₁₃C₆C₉H₂₄O
  • Molecular weight:
    226,31
  • Storage temperature:
    −20 °C
  • Flash point:
    –17 °C - Information refers to the main component,

Specifications

About this item

  • GHS signal: GHS02, GHS08, GHS07
Caution: P201 - P210 - P305 + P351 + P338 - P308 + P313