Supelco® Organic Reference Standard, 4-(3,6-Dimethyl-3-Heptyl)Phenol-3,5-D₂ Diethoxylate Solution
: Merck
- GHS signal: GHS02, GHS07
: P210 - P233 - P240 - P241 - P242 - P305 + P351 + P338
- CAS No.:1173021-42-5
- Storage conditions:–20 °C
- Hazard class description:H225 - H319 - H336
- Shipping conditions:STD
- MDL No.:MFCD09953758
- M-Clarity quality level:MQ100
- Synonym(s):2-{2-[4-(1-Ethyl-1,4-dimethylpentyl)phenoxy-3,5-d₂]ethoxy}ethanol; 2-{2-[4-(3,6-Dimethyl-3-heptyl)phenoxy-3,5-d₂]ethoxy}ethanol; 3,6,3-Nonylphenol diethoxylate-d₂ (ring-3,5-d₂); 363-NP₂EO-D₂
- Formula:C₁₉D₂H₃₀O₃
- Molecular weight:310,47
- Flash point:–17 °C - Information refers to the main component,
- Description:4-(3,6-Dimethyl-3-heptyl)phenol-3,5-d2 diethoxylate solution
Frequently Bought Together