Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, 3-Methylcholanthrene, analytical standard, Supelco®
Organic reference standard, 3-Methylcholanthrene, analytical standard, Supelco®
Catalog # 442388.
Supplier:  Merck
CAS Number:  
Organic reference standard, 3-Methylcholanthrene, analytical standard, Supelco®
Catalog # 442388.
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Standards:
    Analytical standard
  • CAS No.:
    56-49-5
  • Description:
    3-Methylcholanthrene
  • Melting point:
    178...180 °C(lit.)
  • Formula:
    C₂₁H₁₆
  • Storage temperature:
    RT
  • Description:
    3-Methylcholanthrene
  • Boiling point:
    280 °C/80 mmHg(lit.)
  • Pk Info:
    50 mg/ampule

Specifications

About this item

3-Methylcholanthrene, a potent polycyclic aryl hydrocarbon receptor agonist, which is a carcinogen that can be obtained as a byproduct of combustion of various organic substances like coal, garbage, gas and oil.

It suppresses the proliferation of human umbilical vascular endothelial cells.