Order Entry
Austria
ContactUsLinkComponent
Multi-element calibration standard 5
Multi-element calibration standard 5
Catalog # PERCN9300235
Supplier:  PerkinElmer
CAS Number:  
Multi-element calibration standard 5
Catalog # PERCN9300235
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    10 mg/L each
  • Matrix:
    H₂O/tr HF
  • Pk:
    1
  • Description:
    Multi-element calibration standard 5, (12 components)
  • Pk Info:
    125 ml

Specifications

About this item

Designed to contain virtually every element in the mass spectrum for concentration checks.

Concentration of element includes 10 µg/ml of boron, germanium, molybdenum, niobium, phosphorus, rhenium, sulphur, silicon, tantalum, titanium, tungsten, and zirconium.

These solutions are made with the highest purity materials available and are tested on the ELAN 6000 under an internationally accredited ISO 9001 quality assurance program.