Order Entry
Austria
ContactUsLinkComponent
Oil red O solution lipid droplets assay
Flammable
Irritant
Oil red O solution lipid droplets assay
Catalog # CAYM600045-25
Supplier:  Cayman Chemical
CAS Number:  
Flammable
Irritant
Oil red O solution lipid droplets assay
Catalog # CAYM600045-25
Supplier:  Cayman Chemical
CAS Number:  

Specifications

  • Pk:
    25 ml
  • Seq:
    2315

Specifications

About this item

 :
 :408.51 g/mol
Storage Temperature:Refrigerator
UN:1219
ADR:3,II
 :GHS02
 :GHS07
CAS Number:1320-06-5
Formula:C₂₆H₂₄N₄O
MDL Number:MFCD00003898