ContactUsLinkComponent
Dimethylpolysiloxane set of analytical standards, molecular weight series
Dimethylpolysiloxane set of analytical standards, molecular weight series
  361879-1SET
 :  Merck
 :  
undefined
Dimethylpolysiloxane set of analytical standards, molecular weight series
  361879-1SET
 :  Merck
 :  
  • Pk:
    1 SET
  • Description:
    Dimethylpolysiloxane
    set of analytical standards, molecular weight series
Clear, colorless fluid polymer useful as a stationary phase in gas chromatography and as an anti-foaming agent.
It was used in a study performed to understand its molecular weight and molecular weight distribution by off-line coupling of GPC with MALDI mass spectrometer.
Kit components
Mw 40000
Mw 100000
Mw 270000
 : Exact values can be taken from the accompanying certificate of analysis.