Orders LinkContactUsLinkComponent
Stopcocks, three-way
Stopcocks, three-way
Stopcock, three-way
Catalog # SIALCAD6001-1EA
Supplier:   Merck
Stopcocks, three-way
Catalog # SIALCAD6001-1EA
Supplier:  Merck
Stopcocks, three-way
Stopcock, three-way
Price
150,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The spring clip design holds the key in position under proper sealing pressure for optimum performance. There is no need for a screwdriver in cleaning operations.
  • Made from nickel plated brass or stainless steel
  • Tested to 45 psi water pressure
  • High pressure stopcocks are rated to 290 psi
Female luer lock to male luer, right side, female Luer lock, compression-nut.
Specifications
  • Description:
    Stopcock, three-way