

tetra-Chloroauric(III) acid solution 30% (w/v) in diluted hydrochloric acid ≥99.99% (trace metals basis), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:16903-35-8
:
Formula:HAuCl₄
MDL Number:MFCD00011322
:339.78 g/mol
Density:1.637 g/cm³ (25 °C)
UN:3264
ADR:8,II
:M-Clarity™ quality level = MQ100
:GHS05
:GHS07
Specification Test Results
- Appearance (Colour):Yellow to Orange
- Appearance (Form):Liquid
- ICP Major Analysis:Confirmed
- Trace Metal Analysis:≤150.0 ppm
- Purity (Based On Trace Metals Analysis):≤99.99%
Specification Test Results
Frequently Bought Together