ContactUsLinkComponent
Five-fold tap banks, Discofix® C
Five-fold tap banks, Discofix® C
Catalog # BRAU16609C
Supplier:  B. Braun
Five-fold tap banks, Discofix® C
Catalog # BRAU16609C
Supplier:  B. Braun
Specifications
  • Colour:
    Blue
  • Description:
    Five-fold tap bank, drug-resistant
  • Environmentally Preferable:
  • Pk:
    40
Specifications
About this item
Five-fold tap banks are used for administration of medicines and medicines such as anesthetics, cytostatics, antibiotics, narcoleptics, immunosuppressants and lipids.
  • Leaks caused by stress cracks, contamination and air embolism are prevented
  • Easy access through the rotary adapter for injections and aspirations
  • Control of the 45° position by 'clicking'
  • Pressure resistant up to 2 bar
  • PVC and DEHP-free