Orders LinkContactUsLinkComponent
Multi-Purpose Trolleys
Multi-Purpose Trolleys
Supplier:   MULTIROIR
Multi-Purpose Trolleys
Supplier:  MULTIROIR
Certificates
Multi-Purpose Trolleys
6 Options Available Below
Product Details & Documents
These multi-use trolleys have been designed for intensive use. They are made of an aluminium frame with heavy duty polypropylene trays.
  • Grey or black
  • 2 or 3 tray versions
  • Unequalled load capacity: 80 kg per tray and 250 kg per trolley
  • 4 ball bearing wheels diam. 100 mm, 2 with brakes
The trolleys are light, easy to handle and very robust.
Specifications for Product Family
Specifications
  • Catalog No:
  • 216-2232
  • 216-2231
  • 216-2234
  • 216-2236
  • 216-2235
  • 216-2238
  • Model:
  • 65005HS808E3
  • 65007HS808E
  • 65006HS808A3
  • 65001HS808LE3
  • 65003HS808LE
  • 65002HS808LA3
  • Material:
  • PP
  • PP
  • PP
  • PP
  • PP
  • PP
  • Tray dimensions:
  • 615W×450D mm (ext.)
    607W×435D mm (int.)
  • 680W×450D mm (ext.)
    670W×435D mm (int.)
  • 615W×450D mm (ext.)
    607W×435D mm (int.)
  • 920W×520D mm (ext.)
    910W×504D mm (int.)
  • 920W×520D mm (ext.)
    910W×504D mm (int.)
  • 920W×520D mm (ext.)
    910W×504D mm (int.)
  • Height:
  • 553 mm (height between two trays)
  • 553 mm (height between two trays)
  • 275 mm (height between two trays)
  • 553 mm (height between two trays)
  • 553 mm (height between two trays)
  • 275 mm (height between two trays)
Specifications
Product Family Options
Product Information
  • W×D×H
    Shelves
    Colour
    Availability
    Price
  • 800×480×950 mm
    2 trays
    Grey
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 390,00 €
    390,00 €
    1 * 1 ST
       390,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    216-2232
    Model
    65005HS808E3
    Material
    PP
    Tray dimensions
    615W×450D mm (ext.)
    607W×435D mm (int.)
    Height
    553 mm (height between two trays)
  • 800×480×950 mm
    2 trays
    Black
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 377,00 €
    377,00 €
    1 * 1 ST
       377,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    216-2231
    Model
    65007HS808E
    Material
    PP
    Tray dimensions
    680W×450D mm (ext.)
    670W×435D mm (int.)
    Height
    553 mm (height between two trays)
  • 800×480×1000 mm
    3 trays
    Grey
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 454,00 €
    454,00 €
    1 * 1 ST
       454,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    216-2234
    Model
    65006HS808A3
    Material
    PP
    Tray dimensions
    615W×450D mm (ext.)
    607W×435D mm (int.)
    Height
    275 mm (height between two trays)
  • 1100×520×950 mm
    2 trays
    Grey
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 474,00 €
    474,00 €
    1 * 1 ST
       474,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    216-2236
    Model
    65001HS808LE3
    Material
    PP
    Tray dimensions
    920W×520D mm (ext.)
    910W×504D mm (int.)
    Height
    553 mm (height between two trays)
  • 1100×520×950 mm
    2 trays
    Black
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 457,00 €
    457,00 €
    1 * 1 ST
       457,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    216-2235
    Model
    65003HS808LE
    Material
    PP
    Tray dimensions
    920W×520D mm (ext.)
    910W×504D mm (int.)
    Height
    553 mm (height between two trays)
  • 1100×520×1020 mm
    3 trays
    Grey
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 599,00 €
    599,00 €
    1 * 1 ST
       599,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    216-2238
    Model
    65002HS808LA3
    Material
    PP
    Tray dimensions
    920W×520D mm (ext.)
    910W×504D mm (int.)
    Height
    275 mm (height between two trays)