Order Entry
Austria
ContactUsLinkComponent
Tb-color staining kit modified stain for bacteriology, Sigma-Aldrich®
Tb-color staining kit modified stain for bacteriology, Sigma-Aldrich®
Catalog # 1.00497.0001
Supplier:  Merck
CAS Number:  
undefined
Tb-color staining kit modified stain for bacteriology, Sigma-Aldrich®
Catalog # 1.00497.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Fibre carton
  • Pk:
    1
  • Seq:
    0075
  • Pk Info:
    Preparation Capacity 250 slides

Specifications

Specification Test Results

  • Suitability for microscopy:
    passes test
  • Mycobacterium tuberculosis:
    red
  • Background:
    Blue

Specification Test Results

About this item

 :
UN:2924
ADR:3 (8),II
 :GHS02
 :GHS08
 :GHS05
Storage Temperature:Ambient
 :1