TB-fluor fluorescent dye, for bacteriology phenol-free, Sigma-Aldrich®
Catalog # 1.01597.0001
Supplier: Merck
CAS Number:
Specifications
- Pack type:Fibre case
- Pk:1 KIT
- Seq:0075
- Pk Info:Preparation Capacity 60 - 65 slides
Specifications
Specification Test Results
- Suitability for microscopy:passes test
- Mycobacteria:red-orange/yellow-green
- Background:dark
Specification Test Results
About this item
:
UN:3316
ADR:9,II
:GHS02
:GHS07
:GHS09
Storage Temperature:Ambient
:1