Order Entry
Austria
ContactUsLinkComponent
Gram color staining kit modified for Gram staining phenol-free, Sigma-Aldrich®
Gram color staining kit modified for Gram staining phenol-free, Sigma-Aldrich®
Catalog # 1.01603.0001
Supplier:  Merck
CAS Number:  
undefined
Gram color staining kit modified for Gram staining phenol-free, Sigma-Aldrich®
Catalog # 1.01603.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Fibre case
  • Pk:
    1 SET
  • Seq:
    0125
  • Pk Info:
    Preparation Capacity 65 - 70 slides

Specifications

Specification Test Results

  • Suitability for microscopy:
    passes test
  • gram-positve microorganisms:
    dark blue
  • gram-negative microorganisms:
    red

Specification Test Results

About this item

 :
UN:1993
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
Storage Temperature:Ambient
 :1