Order Entry
Austria
ContactUsLinkComponent
GRIESS-ILOSVAY'S nitrite reagent for microbiology, Millipore®
Corrosive
GRIESS-ILOSVAY'S nitrite reagent for microbiology, Millipore®
Catalog # 1.09023.0500
Supplier:  Merck
CAS Number:  
Corrosive
GRIESS-ILOSVAY'S nitrite reagent for microbiology, Millipore®
Catalog # 1.09023.0500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 ml
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    clear, colourless to light pink
  • Sensitivity test:
    passes test

Specification Test Results

About this item

 :
Density:1.03 g/cm3 (20 °C)
Storage Temperature:Ambient
UN:2790
ADR:8,III
 :M-Clarity™ quality level = MQ100
 :GHS05