Order Entry
Austria
ContactUsLinkComponent
Tb-color staining kit stain for bacteriology, Sigma-Aldrich®
Tb-color staining kit stain for bacteriology, Sigma-Aldrich®
Catalog # 1.16450.0001
Supplier:  Merck
CAS Number:  
undefined
Tb-color staining kit stain for bacteriology, Sigma-Aldrich®
Catalog # 1.16450.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Fibre carton
  • Pk:
    1 KIT
  • Seq:
    0075
  • Pk Info:
    4 x 500 ml (Preparation Capacity 250 slides)

Specifications

Specification Test Results

  • Suitability for microscopy:
    passes test
  • Mycobacteria:
    red
  • Background:
    green

Specification Test Results

About this item

 :
ADR:3 (6.1),
 :M-Clarity™ quality level = MQ300
UN:1992
 :GHS02
 :GHS08
 :GHS05
 :GHS09
Storage Temperature:Ambient
 :1