Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Vials
Crimp top vials
Crimp top vials
Crimp top vials
Catalog # 216-3064
Supplier:
RESCH
Expand image
Crimp top vials
Catalog # 216-3064
Supplier:
RESCH
Specifications
Cap included:
No
Ø×H:
27×55 mm
Capacity:
20,00 ml
Pk:
200
Specifications
About this item
Product Details
Crimp top vials made of glass.
Product Details