Order Entry
Austria
ContactUsLinkComponent
Hemacolor® Rapid staining of blood smear solution III staining reagent, Sigma-Aldrich®
Hemacolor® Rapid staining of blood smear solution III staining reagent, Sigma-Aldrich®
Catalog # 1.11957.2500
Supplier:  Merck
CAS Number:  
undefined
Hemacolor® Rapid staining of blood smear solution III staining reagent, Sigma-Aldrich®
Catalog # 1.11957.2500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096
  • Pk Info:
    Preparation Capacity 1250 - 5000 slides

Specifications

About this item

 :
Density:1.01 g/cm3 (20 °C)
Storage Temperature:Ambient
 :1