ContactUsLinkComponent
Poly(ethylene glycol) analytical standards, for GPC, Supelco
Poly(ethylene glycol) analytical standards, for GPC, Supelco
Catalog # 81201-500MG
Supplier:  Merck
CAS Number:  
undefined
Poly(ethylene glycol) analytical standards, for GPC, Supelco
Catalog # 81201-500MG
Supplier:  Merck
CAS Number:  
Specifications
  • Pk:
    500 mg
  • Description:
    ∼96500 Mn, Mw/Mn ~1,12
  • M-Clarity quality level:
    MQ100
Specifications
About this item
Poly(ethylene glycol) is the most widely used non-ionic, hydrophilic polymer, which can find wide applications in biomedical industry such as in drug delivery systems.
Certifications: Exact values can be taken from the accompanying certificate of analysis