Order Entry
Austria
ContactUsLinkComponent
Cell proliferation assay kit, WST-1
Cell proliferation assay kit, WST-1
Catalog # ABNOKA1384
Supplier:  Abnova
CAS Number:  
undefined
Cell proliferation assay kit, WST-1
Catalog # ABNOKA1384
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    WST-1 cell proliferation assay kit

Specifications

About this item

WST-1 cell proliferation assay kit provides an easy to use tool for studying induction and inhibition of cell proliferation in any in vitro model.

  • Application: functional study
  • Store the kit at –20 °C
Caution: For research use only (RUO).