Order Entry
Austria
ContactUsLinkComponent
 
VWR®, Workstation Lab Mat
Catalog # 111-2431
Supplier:  VWR Collection
undefined
VWR®, Workstation Lab Mat
Catalog # 111-2431
Supplier:  VWR Collection

Specifications

  • Colour:
    Green
  • L×W:
    305×379 mm
  • Thickness:
    2 mm
  • Environmentally Preferable:
  • Pk:
    1

Specifications

About this item

These durable FDA food grade silicone lab mats are ideal for every type of laboratory and more specialised areas such as cold rooms, cleanrooms, sterile suites and areas where high sterility is a requirement. They safely facilitate workflow processes with organisation and easy clean up.

  • Reversible - Side one: Combination gridded and smooth surface; Side two: Smooth surface only
  • Keeps bench tops clean and safe from stains, spills and wear, protects against hot items up to +200 °C
  • Lip around the edge can hold approx. 250 ml of liquid
  • Anti-skid surface helps keep the mat and items in place
  • Noise dampening, reducing sound from the vibration of common bench top equipment
  • Chemically inert, does not react with most chemicals
  • Autoclavable at +121 °C for 15 minutes

The gridded surface is ideal for tube staging, preventing movement of small objects and securing items such as reservoirs, pipette tip boxes, assay plates, etc.

These mats are reusable and thus reduce waste making it a more environmentally friendly option over disposable mats. They can be easily cleaned using any standard disinfectant by simply wiping it and are dishwasher safe.

The mats can be rolled up for compact storage.