ContactUsLinkComponent
Giemsa's stain solution for microscopy, Sigma-Aldrich®
Flammable
Toxic
Health
Giemsa's stain solution for microscopy, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Flammable
Toxic
Health
Giemsa's stain solution for microscopy, Sigma-Aldrich®
Supplier:  Merck
Certificates
Specification Test Results
  • Suitability for microscopy:
    passes test
  • Erythrocytes:
    reddish-brownish
  • Nuclei:
    violet
  • Eosinophilic granules:
    reddish to red-brown
  • Neutrophilic granules:
    light violet
  • Lymphocyte cytoplasm:
    Blue
  • Monocyte plasma:
    grey-blue
  • Thrombocytes:
    violet
  • Basophilic granules:
    dark violet
  • Blood parasite nuclei:
    light red
  • Nuclei (Tissue section):
    blue to dark blue
  • Cells:
    blue to dark blue
  • Collagen:
    pale blue
  • Osteoid:
    pale blue
  • Eosinophilic granula (Tissue section):
    red
  • Acidic mucopolysaccharides:
    reddish-violet
  • Mast cell granules:
    reddish-violet
  • Cartilage matrix:
    reddish-violet
  • Acidophilic materials:
    orange-red
  • Helicobacter pylori:
    blue to dark blue