Order Entry
Austria
ContactUsLinkComponent
Croscarmellose sodium PARTECK®, EMPROVE® ESSENTIAL Ph. Eur., JP, NF, SAFC®
Croscarmellose sodium PARTECK®, EMPROVE® ESSENTIAL Ph. Eur., JP, NF, SAFC®
  1.02310.9025
 :  
Croscarmellose sodium PARTECK®, EMPROVE® ESSENTIAL Ph. Eur., JP, NF, SAFC®
  1.02310.9025
 :  

 

  • Pack type:
    Fibre carton
  • Pk:
    25 kg
  • Seq:
    4283

 

 

  • Identity:
    Passes Test
  • Water-Soluble Substances:
    1.0 - 10.0%
  • pH-Value:
    5.0 - 7.0
  • Sodium Chloride and Sodium Glycolate:
    ≤0.5%
  • Heavy Metals (as Pb) (EP):
    ≤20 ppm
  • Heavy Metals (as Pb) (JP):
    ≤10 ppm
  • Heavy Metals (as Pb) (NF):
    ≤10 ppm
  • Ethanol (HS-GC):
    ≤6.0%
  • Other Residual Solvents (ICH Q3C):
    Excluded by the Manufacturing Process
  • Residue on Ignition:
    14.0 - 28.0%
  • Loss on Drying:
    ≤10.0%
  • Degree of Substitution:
    0.60 - 0.85
  • Settling Volume:
    10.0 - 30.0 ml
  • Microbiological Purity (Total Aerobic Microbial Count (TAMC)):
    ≤1000 CFU/g
  • Microbiological Purity (Total Yeast and Mould Count (TYMC)):
    ≤100 CFU/g
  • Escherichia coli (Absent in 1 g):
    Passes Test

 

 

 :
 :982.44 g/mol
 :Ambient
 :74811-65-7
 :C₂₈H₃₀Na₈O₂₇
 :MFCD00217760