Order Entry
Austria
ContactUsLinkComponent
Ethyl acetate ≥99.5% (by GC, corrected for water content), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Ethyl acetate ≥99.5% (by GC, corrected for water content), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # H078-25
   Safer Human and Environmental Health
CAS Number:  
undefined
Ethyl acetate ≥99.5% (by GC, corrected for water content), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # H078-25
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • ACS - Assay (CH₃COOC₂H₅) (by GC, corrected for water):
    ≥99.5 %
  • ACS - Color (APHA):
    ≤10
  • ACS - Residue after Evaporation:
    ≤0.0005 %
  • ACS - Substances Darkened by H₂SO₄:
    Passes Test
  • ACS - Titrable Acid (meq/g):
    ≤0.0009
  • ACS - Water (H₂O)(by Karl Fischer titrn):
    ≤0.05 %
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 255 nm:
    ≤1.00
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 257 nm:
    ≤0.50
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 263 nm:
    ≤0.10
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 265 nm:
    ≤0.05
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 275 nm:
    ≤0.04
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 300 - 330 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 - 400 nm:
    ≤0.005
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 330 - 400 nm:
    ≤0.01

Specification Test Results

About this item

 :
 :88.11 g/mol
Merck Index:13,03792
Boiling Pt:77.1 °C (1013 hPa)
Melting Pt:–83 °C
Density:0.902 g/cm³ (20 °C)
Flash Pt:–4 °C
Storage Temperature:Ambient
EINECS:205-500-4
UN:1173
ADR:3,II
REACH:01-2119475103-46
 :GHS02
 :GHS07
 :GHS08
CAS Number:141-78-6
Formula:CH₃COOC₂H₅
MDL Number:MFCD00009171