ContactUsLinkComponent
Capsule filters, Polycap™ AS, Whatman™
Capsule filters, Polycap™ AS, Whatman™
Certificates
undefined
Capsule filters, Polycap™ AS, Whatman™
Certificates
Specifications
  • Catalog No:
  • 516-0966
  • 516-4445
  • 515-0108
  • 516-0443
  • 516-0957
  • 516-4436
  • 516-4442
  • 516-0965
  • Type:
  • Polycap™ AS 75, nylon, with glass fibre prefilter, non sterile
  • Polycap™ AS 36, nylon, with glass fibre prefilter, sterile
  • Polycap™ AS 36, nylon, with glass fibre prefilter, sterile
  • Polycap™ AS 36, nylon, with glass fibre prefilter, sterile
  • Polycap™ AS 36, nylon, with glass fibre prefilter, non sterile
  • Polycap™ AS 36, nylon, with glass fibre prefilter, sterile
  • Polycap™ AS 36, nylon, with glass fibre prefilter and filling bell, sterile
  • Polycap™ AS 75, nylon, with glass fibre prefilter, non sterile
  • Filtration area:
  • 820 cm²
  • 400 cm²
  • 400 cm²
  • 400 cm²
  • 400 cm²
  • 400 cm²
  • 400 cm²
  • 820 cm²
  • Max. pressure:
  • 4,1 bar
  • 4,1 bar
  • 4,1 bar
  • 4,1 bar
  • 4,1 bar
  • 4,1 bar
  • 4,1 bar
  • 4,1 bar