ContactUsLinkComponent
Multi-element standard, Anions whole volume, certified reference materials (CRMs), for ion chromatography (IC), Supelco®
Multi-element standard, Anions whole volume, certified reference materials (CRMs), for ion chromatography (IC), Supelco®
Catalog # QC3364-500ML
Supplier:  Merck
CAS Number:  
Multi-element standard, Anions whole volume, certified reference materials (CRMs), for ion chromatography (IC), Supelco®
Catalog # QC3364-500ML
Supplier:  Merck
CAS Number:  
Specifications
  • Pk:
    500 mL
  • Description:
    Anions - whole volume
Specifications
About this item
Values of analytes vary lot to lot. Some lots may not contain all analytes..
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.
Caution: Storage temperature: 2 to 8 °C.