Order Entry
Austria
ContactUsLinkComponent
Test paper, qualitative, Watesmo
Test paper, qualitative, Watesmo
Catalog # MANA90609
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Test paper, qualitative, Watesmo
Catalog # MANA90609
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Pk:
    1 Roll
  • Description:
    Water in organic solvents
  • Environmentally Preferable:

Specifications

About this item

The test paper allows the quick and easy detection of water and water vapor.

  • Colour change: Light blue to dark blue

On contact with water the paper changes colour from light blue to dark blue. The reaction is not reversible. Even after drying the contact with water can clearly be proved.

Packaging: Reel of 5 m length and 10 mm width.