Order Entry
Austria
ContactUsLinkComponent
Test strips, qualitative, sulphite
Test strips, qualitative, sulphite
Catalog # MANA90763
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Test strips, qualitative, sulphite
Catalog # MANA90763
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Pk:
    100
  • Description:
    Sulphur dioxide and sulphite ions (SO₂, SO₃²⁻)
  • Range:
    10 mg/l Na₂SO₃

Specifications

About this item

The test paper allows the quick and easy detection of sulphite and sulphur dioxide. In food analysis laboratories it is used to detect sulphite in meat products.

  • Colour changes: White to pink-red

In medical diagnostic it is used as a quick test on sulphite oxidase deficiency.

Packaging: Pack of 100 strips 20×70 mm.