Qualitative test paper peroxtesmo MI for peroxidase in milk
Catalog # MANA90627
Supplier: MACHEREY-NAGEL
CAS Number:
Specifications
- Chemical method:Redox reaction
- Pk:100
- Environmentally Preferable:
- For unit:Peroxtesmo MI test papers, for peroxidase in milk
Specifications
About this item
This test paper allows the specific detection of lactoperoxidase in milk. In the dairy industry the test is used for the quick and easy quality control of highly heated milk. In contrast to the alternative guaiacol method, Peroxtesmo MI is odourless and non-toxic.
- Colour change: White to blue
A drop of milk is put onto the test paper. If the paper remains white, the high heating process is completed.
Limit of sensitivity: 3% raw milk in UHT-milk
Packaging: Pack of 100 test papers 15×15 mm.