Order Entry
Austria
ContactUsLinkComponent
Qualitative test paper peroxtesmo KO for peroxidase in food
Qualitative test paper peroxtesmo KO for peroxidase in food
Catalog # MANA90606
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Qualitative test paper peroxtesmo KO for peroxidase in food
Catalog # MANA90606
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Redox reaction
  • Pk:
    100
  • Description:
    Peroxtesmo KO, for peroxidase in food

Specifications

About this item

This test paper allows the quick and easy detection of peroxidase in food. It is used in the food processing industry to evaluate the quality of canned food.

  • Colour change: White to blue-green

A drop of the sample is put onto the test paper. Alternatively the paper can be pressed to fresh cut surfaces. In the presence of peroxidase a blue colour appears within two minutes.

Packaging: Pack of 100 test papers 15×15 mm.