Order Entry
Austria
Orders LinkContactUsLinkComponent
Test strips, oil

Test strips, oil

Oil Test paper, 250 - mg/L, Wetting of a hydrophobic paper, 48×25×84 mm (package), 100 test(s), 100
Catalog # 711-1990
Supplier:   MACHEREY-NAGEL
Test strips, oil
Catalog # 711-1990
Supplier:  MACHEREY-NAGEL
CAS Number:  
Test strips, oil
Test strips, oil

Oil Test paper, 250 - mg/L, Wetting of a hydrophobic paper, 48×25×84 mm (package), 100 test(s), 100

CAS Number:  
Price
44,90 €
Log in for availability
1 * 100 ST
Product Details & Documents

This test paper allows the rapid and reliable detection of oil contamination of water and soil.

  • Colour changes: Blue to dark blue

For detection of oil in soil, press the paper firmly against the soil to be tested and rinse with clear water. To determine oil in water, move the paper back and forth a few times in the sample.

In case of volatile hydrocarbons, the colour reaction of the test paper has to be evaluated immediately.

The sensitivity largely depends on the nature of the respective hydrocarbon.
Limits of sensitivity:

Petroleum ether: 250 mg/L
Gasoline: 10 mg/L
Fuel oil: 5 mg/L
Lubricating oil: 1 mg/L

Packaging: Pack of 100 strips 20×70 mm.

Specifications

Specifications

  • Chemical method:
    Wetting of a hydrophobic paper
  • Pk:
    100
  • Description:
    Oil Test paper
  • Dimensions:
    48×25×84 mm (package)
  • Tests per kit:
    100 test(s)
  • Range:
    250 - mg/L
  • Cat. No.:
    711-1990
  • Sensitivity:
    Oil - 250 mg/L petroleum ether, 10 mg/L gasoline (high octane), 5 mg/L fuel oil, 1 mg/L lubricating oil
  • Contents:
    100 test strips in a plastic box
  • Platform:
    Qualitative test paper
  • Compliance:
    Not CE certified and no hazardous material
  • Shelf life (months):
    36 (from production)
  • W×D×H:
    20×70 mm (net)
  • Method:
    Oil - pale blue → dark blue
  • Weight:
    49,01 g

Specifications